Learn More
Meso Scale Discovery UPLEX Development Pack6ASY ate
SDP
Catalog No. 5000079993
Encompass
The Meso Scale Discovery U-PLEX Development Pack 6 Assay is a multiplex immunoassay kit that allows users to develop up to 6 custom assays for biomarkers of interest The kit contains plates linkers detection antibodies buffers calibrators and controls needed to develop and run the assays The electrochemiluminescent detection method provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements Assays developed with this kit are compatible with MSD s QuickPlex SQ 120 instrument for rapid results This customizable kit enables researchers to measure panels of biomarkers tailored to their specific needs
Catalog No. 50-000-79993 Supplier Meso Scale Discovery Supplier No. K15231N1
The Meso Scale Discovery U-PLEX Development Pack 6 Assay is a multiplex immunoassay kit that allows users to develop up to 6 custom assays for biomarkers of interest The kit contains plates linkers detection antibodies buffers calibrators and controls needed to develop and run the assays The electrochemiluminescent detection method provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements Assays developed with this kit are compatible with MSD s QuickPlex SQ 120 instrument for rapid results This customizable kit enables researchers to measure panels of biomarkers tailored to their specific needs