Learn More
Meso Scale Discovery UPLEX Development Pack6ASY ate
SDP
Catalog No. 5000082066
Encompass
The Meso Scale Discovery U-PLEX Development Pack 6 Assay is a multiplex immunoassay kit that allows users to develop up to 6 custom assays for biomarkers of interest The kit contains plates linkers detection antibodies buffers calibrators and controls needed to develop and run the assays The electrochemiluminescent detection method provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements Assays developed with this kit are compatible with MSD s QuickPlex SQ 120 instrument for rapid results Overall this is a customizable kit that enables sensitive multiplex immunoassays tailored to specific research needs
Catalog No. 50-000-82066 Supplier Meso Scale Discovery Supplier No. K15231N4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Development Pack 6 Assay is a multiplex immunoassay kit that allows users to develop up to 6 custom assays for biomarkers of interest The kit contains plates linkers detection antibodies buffers calibrators and controls needed to develop and run the assays The electrochemiluminescent detection method provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements Assays developed with this kit are compatible with MSD s QuickPlex SQ 120 instrument for rapid results Overall this is a customizable kit that enables sensitive multiplex immunoassays tailored to specific research needs