Learn More
Meso Scale Discovery UPLEX Development Pack7ASY ate
SDP
Catalog No. 5000079250
Encompass
The Meso Scale Discovery U-PLEX Development Pack 7 Assay is a multiplex immunoassay kit for developing custom assays for up to 7 biomarkers It includes a 10-plate MULTI-ARRAY 96-well small-spot plate SULFO-TAG NHS-Ester labels Read Buffer Sample Diluent Wash Buffer Blocker A and detection antibodies This kit supports exploratory research in immunology oncology and neurobiology with electrochemiluminescence technology offering high sensitivity a wide dynamic range and low sample volume requirements It provides reagents for developing and optimizing custom multiplex immunoassays
Catalog No. 50-000-79250 Supplier Meso Scale Discovery Supplier No. K15232N4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Development Pack 7 Assay is a multiplex immunoassay kit for developing custom assays for up to 7 biomarkers It includes a 10-plate MULTI-ARRAY 96-well small-spot plate SULFO-TAG NHS-Ester labels Read Buffer Sample Diluent Wash Buffer Blocker A and detection antibodies This kit supports exploratory research in immunology oncology and neurobiology with electrochemiluminescence technology offering high sensitivity a wide dynamic range and low sample volume requirements It provides reagents for developing and optimizing custom multiplex immunoassays