Learn More
Meso Scale Discovery UPLEX Development Pack7ASY ate
SDP
Catalog No. 5000081404
Encompass
The Meso Scale Discovery U-PLEX Development Pack 7 Assay is a multiplex immunoassay kit that allows users to develop custom assays for measuring up to 7 biomarkers simultaneously from a single sample The kit contains a 10-plate MULTI-ARRAY 96-well plate reagents and buffers needed to develop and optimize custom assays using electrochemiluminescence detection on the MESO QuickPlex SQ 120 instrument The kit enables quantification of protein biomarkers antibodies and other analytes in biological samples It provides reagents to develop optimize and validate custom multiplex immunoassays
Catalog No. 50-000-81404 Supplier Meso Scale Discovery Supplier No. K15232N1
The Meso Scale Discovery U-PLEX Development Pack 7 Assay is a multiplex immunoassay kit that allows users to develop custom assays for measuring up to 7 biomarkers simultaneously from a single sample The kit contains a 10-plate MULTI-ARRAY 96-well plate reagents and buffers needed to develop and optimize custom assays using electrochemiluminescence detection on the MESO QuickPlex SQ 120 instrument The kit enables quantification of protein biomarkers antibodies and other analytes in biological samples It provides reagents to develop optimize and validate custom multiplex immunoassays