Learn More
Meso Scale Discovery UPLEX Development Pack8ASY ate
SDP
Catalog No. 5000080014
Encompass
The Meso Scale Discovery U-PLEX Development Pack 8 Assay K15233N is a multiplex immunoassay kit that allows users to develop up to 8 custom assays for biomarker detection The kit contains plates linkers detection antibodies buffers calibrators and controls needed to develop and optimize custom U-PLEX assays The electrochemiluminescent detection method provides high sensitivity up to 5 logs of dynamic range and low sample consumption This makes the kit useful for developing custom biomarker panels for cancer immunology neurobiology and biomarker research
Catalog No. 50-000-80014 Supplier Meso Scale Discovery Supplier No. K15233N4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Development Pack 8 Assay K15233N is a multiplex immunoassay kit that allows users to develop up to 8 custom assays for biomarker detection The kit contains plates linkers detection antibodies buffers calibrators and controls needed to develop and optimize custom U-PLEX assays The electrochemiluminescent detection method provides high sensitivity up to 5 logs of dynamic range and low sample consumption This makes the kit useful for developing custom biomarker panels for cancer immunology neurobiology and biomarker research