Learn More
Meso Scale Discovery UPLEX Development Pack8ASY ate
SDP
Catalog No. 5000082356
Encompass
The Meso Scale Discovery U-PLEX Development Pack 8 Assay is a multiplex immunoassay kit that allows users to develop up to 8 custom assays for biomarkers of interest The kit contains plates linkers detection antibodies buffers calibrators and controls needed to develop and optimize custom assays The electrochemiluminescent detection method provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements Assay development time is reduced compared to traditional ELISA This product enables custom multiplex protein biomarker analysis for life science research applications
Catalog No. 50-000-82356 Supplier Meso Scale Discovery Supplier No. K15233N1
The Meso Scale Discovery U-PLEX Development Pack 8 Assay is a multiplex immunoassay kit that allows users to develop up to 8 custom assays for biomarkers of interest The kit contains plates linkers detection antibodies buffers calibrators and controls needed to develop and optimize custom assays The electrochemiluminescent detection method provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements Assay development time is reduced compared to traditional ELISA This product enables custom multiplex protein biomarker analysis for life science research applications