Learn More
Meso Scale Discovery UPLEX Development Pack9ASY ate
SDP
Catalog No. 5000081352
Encompass
The Meso Scale Discovery U-PLEX Development Pack 9 Assay is a multiplex immunoassay kit that allows users to develop custom assays for up to 9 biomarkers It contains the components needed to develop and optimize an assay including plates linkers detection antibodies buffers calibrators and controls The electrochemiluminescent detection method provides high sensitivity up to 5 logs of dynamic range and low sample consumption This makes it useful for biomarker discovery and validation The kit allows flexibility in assay development while utilizing MSD s sensitive MULTI-ARRAY technology
Catalog No. 50-000-81352 Supplier Meso Scale Discovery Supplier No. K15234N2
The Meso Scale Discovery U-PLEX Development Pack 9 Assay is a multiplex immunoassay kit that allows users to develop custom assays for up to 9 biomarkers It contains the components needed to develop and optimize an assay including plates linkers detection antibodies buffers calibrators and controls The electrochemiluminescent detection method provides high sensitivity up to 5 logs of dynamic range and low sample consumption This makes it useful for biomarker discovery and validation The kit allows flexibility in assay development while utilizing MSD s sensitive MULTI-ARRAY technology