Learn More
Meso Scale Discovery UPLEX EffectorCell Checkpoint
SDP
Catalog No. 5000082048
Encompass
The U-PLEX Effector Cell Checkpoint Combo 1 Human Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous measurement of human PD-1 PD-L1 CTLA-4 LAG-3 TIM-3 and TIGIT from cell culture supernatants serum plasma and other biological samples The kit contains a 96-well plate coated with capture antibodies detection antibodies and calibrators to quantify each analyte The electrochemiluminescent detection allows high sensitivity and a wide dynamic range This kit enables efficient profiling of checkpoint molecules on effector T cells and NK cells involved in immune response regulation
Catalog No. 50-000-82048 Supplier Meso Scale Discovery Supplier No. K15341K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Effector Cell Checkpoint Combo 1 Human Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows simultaneous measurement of human PD-1 PD-L1 CTLA-4 LAG-3 TIM-3 and TIGIT from cell culture supernatants serum plasma and other biological samples The kit contains a 96-well plate coated with capture antibodies detection antibodies and calibrators to quantify each analyte The electrochemiluminescent detection allows high sensitivity and a wide dynamic range This kit enables efficient profiling of checkpoint molecules on effector T cells and NK cells involved in immune response regulation