Learn More
Meso Scale Discovery UPLEX EffectorCell Checkpoint
SDP
Catalog No. 5000079390
Encompass
The U-PLEX Effector Cell Checkpoint Combo 1 Human Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous measurement of human IFN-gamma TNF-alpha IL-2 IL-4 IL-6 IL-10 IL-17A IL-17F IL-21 IL-22 IL-23 IL-31 sCD40L and GM-CSF from cell culture supernatants serum plasma and other biological samples The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and wash buffer The electrochemiluminescent assay allows for high sensitivity and a broad dynamic range
Catalog No. 50-000-79390 Supplier Meso Scale Discovery Supplier No. K15341K2
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Effector Cell Checkpoint Combo 1 Human Kit from Meso Scale Discovery is a multiplex immunoassay kit that allows for the simultaneous measurement of human IFN-gamma TNF-alpha IL-2 IL-4 IL-6 IL-10 IL-17A IL-17F IL-21 IL-22 IL-23 IL-31 sCD40L and GM-CSF from cell culture supernatants serum plasma and other biological samples The kit contains a 96-well plate coated with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and wash buffer The electrochemiluminescent assay allows for high sensitivity and a broad dynamic range