Learn More
Meso Scale Discovery UPLEX EffectorCell Checkpoint
SDP
Catalog No. 5000079844
Encompass
The U-PLEX Effector Cell Checkpoint Combo 1 Human Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring human IFN-gamma TNF-alpha Granzyme B Perforin IL-2 IL-4 IL-6 IL-10 IL-17A IL-17F IL-21 IL-22 and IL-31 in various biological matrices It includes a 96-well plate with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The kit quantifies effector cell functions and cytokines related to checkpoint inhibition therapy offering high sensitivity low sample volume and reproducible results
Catalog No. 50-000-79844 Supplier Meso Scale Discovery Supplier No. K15341K1
The U-PLEX Effector Cell Checkpoint Combo 1 Human Kit from Meso Scale Discovery is a multiplex immunoassay kit for measuring human IFN-gamma TNF-alpha Granzyme B Perforin IL-2 IL-4 IL-6 IL-10 IL-17A IL-17F IL-21 IL-22 and IL-31 in various biological matrices It includes a 96-well plate with capture antibodies detection antibodies Read Buffer Diluent 2 Calibrator Blend and Wash Buffer The kit quantifies effector cell functions and cytokines related to checkpoint inhibition therapy offering high sensitivity low sample volume and reproducible results