Learn More
Meso Scale Discovery UPLEX Gut Hormone Combo 1 ms
SDP
Catalog No. 5000079114
Encompass
The Meso Scale Discovery U-PLEX Gut Hormone Combo 1 Mouse Kit is a multiplex immunoassay for measuring active ghrelin leptin PYY and GLP-1 in mouse samples It includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and plate seals Using electrochemiluminescence technology it offers high sensitivity and a broad dynamic range Suitable for studying appetite regulation and metabolic diseases the kit requires 50 L of sample and is compatible with cell culture supernatants serum plasma and tissue lysates
Catalog No. 50-000-79114 Supplier Meso Scale Discovery Supplier No. K15307K1
The Meso Scale Discovery U-PLEX Gut Hormone Combo 1 Mouse Kit is a multiplex immunoassay for measuring active ghrelin leptin PYY and GLP-1 in mouse samples It includes a 96-well plate with capture antibodies detection antibodies calibrators controls diluent read buffer wash buffer and plate seals Using electrochemiluminescence technology it offers high sensitivity and a broad dynamic range Suitable for studying appetite regulation and metabolic diseases the kit requires 50 L of sample and is compatible with cell culture supernatants serum plasma and tissue lysates