Learn More
Meso Scale Discovery UPLEX Gut Hormone Combo 1 ms
SDP
Catalog No. 5000080417
Encompass
The Meso Scale Discovery U-PLEX Gut Hormone Combo 1 Mouse Kit is a multiplex immunoassay that measures CCK GIP GLP-1 and PYY in mouse samples This assay kit allows for the simultaneous detection of these 4 gut hormones from a single sample It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volumes The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents needed to run the assay It can be used to study gut hormone biology and their role in metabolism and appetite regulation in mouse models
Catalog No. 50-000-80417 Supplier Meso Scale Discovery Supplier No. K15307K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Gut Hormone Combo 1 Mouse Kit is a multiplex immunoassay that measures CCK GIP GLP-1 and PYY in mouse samples This assay kit allows for the simultaneous detection of these 4 gut hormones from a single sample It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volumes The kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators and reagents needed to run the assay It can be used to study gut hormone biology and their role in metabolism and appetite regulation in mouse models