Learn More
Meso Scale Discovery UPLEX Gut Hormone Combo 1 ms
SDP
Catalog No. 5000082393
Encompass
The Meso Scale Discovery U-PLEX Gut Hormone Combo 1 Mouse Kit is a multiplex immunoassay that allows simultaneous measurement of active ghrelin leptin PYY and GLP-1 in mouse samples This kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals The assay has a typical sensitivity of 0 3-2 4 pg/mL and requires 25 L of sample It utilizes electrochemiluminescence detection and can be read on MSD instrument platforms This kit allows efficient quantification of multiple gut hormones from mouse samples
Catalog No. 50-000-82393 Supplier Meso Scale Discovery Supplier No. K15307K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Gut Hormone Combo 1 Mouse Kit is a multiplex immunoassay that allows simultaneous measurement of active ghrelin leptin PYY and GLP-1 in mouse samples This kit includes a 96-well plate coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer wash buffer and plate seals The assay has a typical sensitivity of 0 3-2 4 pg/mL and requires 25 L of sample It utilizes electrochemiluminescence detection and can be read on MSD instrument platforms This kit allows efficient quantification of multiple gut hormones from mouse samples