Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meso Scale Discovery U-PLEX Human gp130 (soluble) Assay, Single-analyte, 384-well plate format
SDP

Catalog No. NC3315355
Encompass

The MESO SCALE DISCOVERY U-PLEX Human gp130 (soluble) Assay is a highly sensitive, single-analyte immunoassay designed for the quantitative measurement of soluble human gp130 in various biological samples. Utilizing electrochemiluminescence (ECL) detection technology, this assay is engineered for ease of use, enabling researchers to create high-quality assays efficiently. Suitable for research applications, the U-PLEX platform enhances multipurpose capabilities by allowing for potential multiplexing with other analytes.

  • Analyte: Human gp130 (soluble)
  • Assay format: Single-analyte
  • Detection method: Electrochemiluminescence (ECL)
  • Plate format: 384-well
  • Application: Research use only
  • Sample types: Serum, plasma, cell culture supernatant
  • Multiplexing capability: Yes
  • Throughput: High

Catalog No. NC3315355 Supplier Meso Scale Discovery Supplier No. K151P7K2
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The MESO SCALE DISCOVERY U-PLEX Human gp130 (soluble) Assay is a highly sensitive, single-analyte immunoassay designed for the quantitative measurement of soluble human gp130 in various biological samples. Utilizing electrochemiluminescence (ECL) detection technology, this assay is engineered for ease of use, enabling researchers to create high-quality assays efficiently. Suitable for research applications, the U-PLEX platform enhances multipurpose capabilities by allowing for potential multiplexing with other analytes.

  • Analyte: Human gp130 (soluble)
  • Assay format: Single-analyte
  • Detection method: Electrochemiluminescence (ECL)
  • Plate format: 384-well
  • Application: Research use only
  • Sample types: Serum, plasma, cell culture supernatant
  • Multiplexing capability: Yes
  • Throughput: High

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.