Learn More
Meso Scale Discovery UPLEX HUM A2M ASY384well
SDP
Catalog No. 5000079861
Encompass
The Meso Scale Discovery U-PLEX Human A2M 384-Well Assay is a multiplex immunoassay panel that measures Alpha-2-Macroglobulin in human serum plasma and cell culture supernatant samples This assay utilizes electrochemiluminescence detection technology and is designed for use on Meso Scale Discovery instrumentation including the QuickPlex SQ 120 QuickPlex SQ 120i MESO QuickPlex SQ 120 and QuickPlex SQ 120 The assay can process up to 384 samples at once and has a testing time of approximately 3 5 hours The quantitative range of the assay is between 50 - 50 000 pg/mL This product requires the separate purchase of a Meso Scale Discovery instrument to run the assay
Catalog No. 50-000-79861 Supplier Meso Scale Discovery Supplier No. K251Q9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human A2M 384-Well Assay is a multiplex immunoassay panel that measures Alpha-2-Macroglobulin in human serum plasma and cell culture supernatant samples This assay utilizes electrochemiluminescence detection technology and is designed for use on Meso Scale Discovery instrumentation including the QuickPlex SQ 120 QuickPlex SQ 120i MESO QuickPlex SQ 120 and QuickPlex SQ 120 The assay can process up to 384 samples at once and has a testing time of approximately 3 5 hours The quantitative range of the assay is between 50 - 50 000 pg/mL This product requires the separate purchase of a Meso Scale Discovery instrument to run the assay