Learn More
Meso Scale Discovery UPLEX HUM Adiponectin ASY 384
SDP
Catalog No. 5000080680
Encompass
The Meso Scale Discovery U-PLEX Human Adiponectin 384-Well Assay Kit is a multiplex immunoassay that measures human adiponectin levels in serum plasma and cell culture supernatants This kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay for high throughput analysis of adiponectin with high sensitivity and broad dynamic range The assay requires a low sample volume of 25 L and has a testing time of approximately 3 5 hours This product can aid in understanding adiponectin biology and the role of adiponectin in obesity diabetes cardiovascular disease and other disorders
Catalog No. 50-000-80680 Supplier Meso Scale Discovery Supplier No. K251R9K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Adiponectin 384-Well Assay Kit is a multiplex immunoassay that measures human adiponectin levels in serum plasma and cell culture supernatants This kit uses electrochemiluminescence detection technology and provides a 384-well plate formatted assay for high throughput analysis of adiponectin with high sensitivity and broad dynamic range The assay requires a low sample volume of 25 L and has a testing time of approximately 3 5 hours This product can aid in understanding adiponectin biology and the role of adiponectin in obesity diabetes cardiovascular disease and other disorders