Learn More
Meso Scale Discovery UPLEX HUM Adiponectin ASY 384
SDP
Catalog No. 5000081065
Encompass
The Meso Scale Discovery U-PLEX Human Adiponectin 384-Well Assay Kit is a multiplex immunoassay that measures human adiponectin levels in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 384-well plate coated with capture antibodies detection antibodies calibrators and controls It requires a small sample volume of 25 L and the total assay time is around 3 hours The kit can quantify adiponectin levels from 50 pg/mL to 10 000 pg/mL
Catalog No. 50-000-81065 Supplier Meso Scale Discovery Supplier No. K251R9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Adiponectin 384-Well Assay Kit is a multiplex immunoassay that measures human adiponectin levels in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results The kit includes a 384-well plate coated with capture antibodies detection antibodies calibrators and controls It requires a small sample volume of 25 L and the total assay time is around 3 hours The kit can quantify adiponectin levels from 50 pg/mL to 10 000 pg/mL