Learn More
Meso Scale Discovery UPLEX HUM Adiponectin ASY QP
SDP
Catalog No. 5000079221
Encompass
The Meso Scale Discovery U-PLEX Human Adiponectin Assay Kit is designed for the quantitative measurement of human adiponectin in serum plasma cell culture supernatants and other biological samples It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The assay has high specificity for human adiponectin with no cross-reactivity to other proteins It requires 2 5 hours to complete and includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer diluent and calibrators The kit can perform up to 25 tests for human adiponectin quantification
Catalog No. 50-000-79221 Supplier Meso Scale Discovery Supplier No. K151R9K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Adiponectin Assay Kit is designed for the quantitative measurement of human adiponectin in serum plasma cell culture supernatants and other biological samples It utilizes MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity a wide dynamic range and requires low sample volume The assay has high specificity for human adiponectin with no cross-reactivity to other proteins It requires 2 5 hours to complete and includes a 96-well plate coated with capture antibody detection antibody read buffer wash buffer diluent and calibrators The kit can perform up to 25 tests for human adiponectin quantification