Learn More
Meso Scale Discovery UPLEX HUM Adiponectin ASY QP
SDP
Catalog No. 5000080589
Encompass
The Meso Scale Discovery U-PLEX Human Adiponectin Assay Kit is designed for the quantitative measurement of adiponectin in human serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses a plate coated with capture antibodies and U-PLEX linker conjugated detection antibodies specific for adiponectin It has a detection range of 50-10 000 pg/mL and includes calibrators and controls The kit includes a 96-well plate detection antibody read buffer wash buffer sample diluent and complete instructions It provides high sensitivity and broad dynamic range for adiponectin quantification
Catalog No. 50-000-80589 Supplier Meso Scale Discovery Supplier No. K151R9K22
The Meso Scale Discovery U-PLEX Human Adiponectin Assay Kit is designed for the quantitative measurement of adiponectin in human serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses a plate coated with capture antibodies and U-PLEX linker conjugated detection antibodies specific for adiponectin It has a detection range of 50-10 000 pg/mL and includes calibrators and controls The kit includes a 96-well plate detection antibody read buffer wash buffer sample diluent and complete instructions It provides high sensitivity and broad dynamic range for adiponectin quantification