Learn More
Meso Scale Discovery UPLEX HUM Adiponectin ASY
SDP
Catalog No. 5000079222
Encompass
The Meso Scale Discovery U-PLEX Human Adiponectin Assay Kit is designed for the quantitative measurement of human adiponectin in serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses a plate-based format and provides high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags and calibrator diluent The assay takes approximately 3 5 hours to complete
Catalog No. 50-000-79222 Supplier Meso Scale Discovery Supplier No. K151R9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Adiponectin Assay Kit is designed for the quantitative measurement of human adiponectin in serum plasma and cell culture supernatants using an electrochemiluminescent detection method The assay uses a plate-based format and provides high sensitivity a broad dynamic range and requires a low sample volume The kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags and calibrator diluent The assay takes approximately 3 5 hours to complete