Learn More
Meso Scale Discovery UPLEX HUM ApoA1 ASY384well
SDP
Catalog No. 5000079569
Encompass
The Meso Scale Discovery U-PLEX Human ApoA1 384-Well Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of Apolipoprotein A1 (ApoA1) in human serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and buffers for 384 tests The assay has a typical sensitivity of 0 3 ng/mL and a detection range of 0 3-2500 ng/mL Results are obtained in as little as 3 5 hours using Meso Scale Discovery s instrumentation for ECL detection This assay allows high-throughput sensitive and accurate quantification of ApoA1
Catalog No. 50-000-79569 Supplier Meso Scale Discovery Supplier No. K251S9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human ApoA1 384-Well Assay Kit is an electrochemiluminescent immunoassay for the quantitative measurement of Apolipoprotein A1 (ApoA1) in human serum plasma and cell culture supernatants The kit contains a 384-well plate coated with capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and buffers for 384 tests The assay has a typical sensitivity of 0 3 ng/mL and a detection range of 0 3-2500 ng/mL Results are obtained in as little as 3 5 hours using Meso Scale Discovery s instrumentation for ECL detection This assay allows high-throughput sensitive and accurate quantification of ApoA1