Learn More
Meso Scale Discovery UPLEX HUM ApoC3 ASY384well
SDP
Catalog No. 5000080685
Encompass
The Meso Scale Discovery U-PLEX Human ApoC3 384-Well Assay is a multiplex immunoassay kit for measuring Apolipoprotein C3 (ApoC3) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs The assay requires 10 L per well and can perform up to 40 tests on two plates The kit includes a 384-well plate with capture antibodies detection antibodies calibrators and controls with a detection range of 17 to 100 000 pg/mL for ApoC3
Catalog No. 50-000-80685 Supplier Meso Scale Discovery Supplier No. K251T9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human ApoC3 384-Well Assay is a multiplex immunoassay kit for measuring Apolipoprotein C3 (ApoC3) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a 5-log dynamic range and low sample volume needs The assay requires 10 L per well and can perform up to 40 tests on two plates The kit includes a 384-well plate with capture antibodies detection antibodies calibrators and controls with a detection range of 17 to 100 000 pg/mL for ApoC3