Learn More
Meso Scale Discovery UPLEX HUM ApoC3 ASYQP 25 PL
SDP
Catalog No. 5000080546
Encompass
The Meso Scale Discovery U-PLEX Human ApoC3 Assay Kit quantitatively measures Apolipoprotein C3 (ApoC3) in human serum plasma and cell culture supernatants This electrochemiluminescent multiplex immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection of ApoC3 levels down to the pg/mL range It requires a small sample volume and offers high-throughput capabilities for research on lipid metabolism cardiovascular disease diabetes and metabolic disorders The kit includes a 96-well plate with capture antibody detection antibody read buffer calibrator and sample diluent
Catalog No. 50-000-80546 Supplier Meso Scale Discovery Supplier No. K151T9K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human ApoC3 Assay Kit quantitatively measures Apolipoprotein C3 (ApoC3) in human serum plasma and cell culture supernatants This electrochemiluminescent multiplex immunoassay uses MSD s SULFO-TAG labels and multi-array technology for sensitive detection of ApoC3 levels down to the pg/mL range It requires a small sample volume and offers high-throughput capabilities for research on lipid metabolism cardiovascular disease diabetes and metabolic disorders The kit includes a 96-well plate with capture antibody detection antibody read buffer calibrator and sample diluent