Learn More
Meso Scale Discovery UPLEX HUM APRIL/TNFSF13 ASY
SDP
Catalog No. 5000081583
Encompass
The Meso Scale Discovery U-PLEX Human APRIL (TNFSF13) Assay is a multiplex immunoassay kit that enables simultaneous quantification of APRIL in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and plate seals It allows sensitive reproducible and rapid measurement of APRIL levels within 3 5 hours
Catalog No. 50-000-81583 Supplier Meso Scale Discovery Supplier No. K151AGUK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human APRIL (TNFSF13) Assay is a multiplex immunoassay kit that enables simultaneous quantification of APRIL in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies calibrators controls 10X diluent read buffer and plate seals It allows sensitive reproducible and rapid measurement of APRIL levels within 3 5 hours