Learn More
Meso Scale Discovery UPLEX HUM APRIL/TNFSF13 ASY384
SDP
Catalog No. 5000081115
Encompass
The Meso Scale Discovery U-PLEX Human APRIL (TNFSF13) Assay is a 384-well electrochemiluminescent immunoassay for measuring APRIL levels in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive quantitation of APRIL with a quantification range of 19 5 to 10 000 pg/mL Requiring only 25 L of sample it can be read on MSD s SECTOR Imager 6000 facilitating high throughput screening This assay reliably measures APRIL crucial for immune function and autoimmune disease research
Catalog No. 50-000-81115 Supplier Meso Scale Discovery Supplier No. K251AGUK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human APRIL (TNFSF13) Assay is a 384-well electrochemiluminescent immunoassay for measuring APRIL levels in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array plate technology for sensitive quantitation of APRIL with a quantification range of 19 5 to 10 000 pg/mL Requiring only 25 L of sample it can be read on MSD s SECTOR Imager 6000 facilitating high throughput screening This assay reliably measures APRIL crucial for immune function and autoimmune disease research