Learn More
Meso Scale Discovery UPLEX HUM APRIL/TNFSF13 ASYQP
SDP
Catalog No. 5000080592
Encompass
The Meso Scale Discovery U-PLEX Human APRIL (TNFSF13) Assay is a multiplex immunoassay kit for measuring APRIL (TNFSF13) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity up to 5 logs of dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls for accurate APRIL measurement It provides a rapid reproducible method for quantifying APRIL a TNF superfamily member involved in immune regulation
Catalog No. 50-000-80592 Supplier Meso Scale Discovery Supplier No. K151AGUK22
The Meso Scale Discovery U-PLEX Human APRIL (TNFSF13) Assay is a multiplex immunoassay kit for measuring APRIL (TNFSF13) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity up to 5 logs of dynamic range and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies and necessary calibrators and controls for accurate APRIL measurement It provides a rapid reproducible method for quantifying APRIL a TNF superfamily member involved in immune regulation