Learn More
Meso Scale Discovery UPLEX HUM APRIL/TNFSF13 ASYQP
SDP
Catalog No. 5000081574
Encompass
The Meso Scale Discovery U-PLEX Human APRIL (TNFSF13) Assay is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of APRIL (TNFSF13) in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure APRIL levels accurately It provides a rapid reproducible method for quantifying APRIL a cytokine involved in immune regulation
Catalog No. 50-000-81574 Supplier Meso Scale Discovery Supplier No. K151AGUK24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human APRIL (TNFSF13) Assay is a multiplex immunoassay kit that allows for the simultaneous quantitative measurement of APRIL (TNFSF13) in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate pre-coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure APRIL levels accurately It provides a rapid reproducible method for quantifying APRIL a cytokine involved in immune regulation