Learn More
Meso Scale Discovery UPLEX HUM APRIL/TNFSF1ASY384
SDP
Catalog No. 5000080985
Encompass
The Meso Scale Discovery U-PLEX Human APRIL (TNFSF13) Assay is a 384-well electrochemiluminescent immunoassay designed to measure APRIL levels in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible quantitation of APRIL with a broad dynamic range The kit includes a 384-well plate coated with an APRIL capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and buffers for sample dilution The assay has a lower limit of detection of 0 04 ng/mL and an upper limit of 10 000 pg/mL
Catalog No. 50-000-80985 Supplier Meso Scale Discovery Supplier No. K251AGUK2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human APRIL (TNFSF13) Assay is a 384-well electrochemiluminescent immunoassay designed to measure APRIL levels in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to provide sensitive and reproducible quantitation of APRIL with a broad dynamic range The kit includes a 384-well plate coated with an APRIL capture antibody detection antibody conjugated to an electrochemiluminescent label calibrators and buffers for sample dilution The assay has a lower limit of detection of 0 04 ng/mL and an upper limit of 10 000 pg/mL