Learn More
Meso Scale Discovery UPLEX HUM BAF/TNFRSF13C ASY384
SDP
Catalog No. 5000079567
Encompass
The Meso Scale Discovery U-PLEX Human BAFF/TNFRSF13C 384-well Assay Kit is a multiplex immunoassay that measures human BAFF and TNFRSF13C in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and controls It allows simultaneous measurement of BAFF and TNFRSF13C from a 50 L sample in 4 5 hours
Catalog No. 50-000-79567 Supplier Meso Scale Discovery Supplier No. K251K7K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human BAFF/TNFRSF13C 384-well Assay Kit is a multiplex immunoassay that measures human BAFF and TNFRSF13C in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and intra- and inter-assay precision The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and controls It allows simultaneous measurement of BAFF and TNFRSF13C from a 50 L sample in 4 5 hours