Learn More
Meso Scale Discovery UPLEX HUM BAF/TNFRSF13C ASY384
SDP
Catalog No. 5000080697
Encompass
The Meso Scale Discovery U-PLEX Human BAFF/TNFRSF13C 384-Well Assay Kit is a multiplex immunoassay that measures human BAFF and TNFRSF13C in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and controls It allows simultaneous measurement of BAFF and TNFRSF13C from a single small sample volume The kit has high performance and reliability for biomarker research and drug development applications
Catalog No. 50-000-80697 Supplier Meso Scale Discovery Supplier No. K251K7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human BAFF/TNFRSF13C 384-Well Assay Kit is a multiplex immunoassay that measures human BAFF and TNFRSF13C in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent labels calibrators and controls It allows simultaneous measurement of BAFF and TNFRSF13C from a single small sample volume The kit has high performance and reliability for biomarker research and drug development applications