Learn More
Meso Scale Discovery UPLEX HUM BAF/TNFRSF13C ASYQP
SDP
Catalog No. 5000082346
Encompass
The Meso Scale Discovery U-PLEX Human BAFF/TNFRSF13C Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of BAFF and TNFRSF13C in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on MSD s SECTOR Imager 6000 It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure BAFF and TNFRSF13C concentrations
Catalog No. 50-000-82346 Supplier Meso Scale Discovery Supplier No. K151K7K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human BAFF/TNFRSF13C Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of BAFF and TNFRSF13C in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on MSD s SECTOR Imager 6000 It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure BAFF and TNFRSF13C concentrations