Learn More
Meso Scale Discovery UPLEX HUM BAF/TNFRSF13C ASYQP
SDP
Catalog No. 5000079547
Encompass
The Meso Scale Discovery U-PLEX Human BAFF/TNFRSF13C Assay is a multiplex immunoassay kit that enables simultaneous quantification of BAFF and TNFRSF13C in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on MSD s SECTOR Imager 6000 It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure sample concentrations
Catalog No. 50-000-79547 Supplier Meso Scale Discovery Supplier No. K151K7K21
The Meso Scale Discovery U-PLEX Human BAFF/TNFRSF13C Assay is a multiplex immunoassay kit that enables simultaneous quantification of BAFF and TNFRSF13C in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements The assay requires 50 L of sample and can be read on MSD s SECTOR Imager 6000 It includes a plate coated with capture antibodies detection antibodies and calibrators and controls needed to generate a standard curve and measure sample concentrations