Learn More
Meso Scale Discovery UPLEX HUM BAF/TNFRSF13C ASYQP
SDP
Catalog No. 5000082345
Encompass
The Meso Scale Discovery U-PLEX Human BAFF/TNFRSF13C Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of BAFF and TNFRSF13C in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent It can monitor BAFF and TNFRSF13C levels and has research applications in immunology and oncology studies
Catalog No. 50-000-82345 Supplier Meso Scale Discovery Supplier No. K151K7K22
The Meso Scale Discovery U-PLEX Human BAFF/TNFRSF13C Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of BAFF and TNFRSF13C in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent It can monitor BAFF and TNFRSF13C levels and has research applications in immunology and oncology studies