Learn More
Meso Scale Discovery UPLEX HUM BAFF ASY384well
SDP
Catalog No. 5000080779
Encompass
The Meso Scale Discovery U-PLEX BAFF Singleplex Assay Kit is a highly sensitive and quantitative electrochemiluminescent immunoassay for the detection of B cell activating factor (BAFF) in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to enable rapid and accurate measurement of BAFF levels with high sensitivity and a broad dynamic range The assay requires only 50 L of sample and can be read on MSD s SECTOR Imager instruments This assay provides a simple rapid and robust tool for investigating BAFF biology and evaluating BAFF-targeted therapeutics
Catalog No. 50-000-80779 Supplier Meso Scale Discovery Supplier No. K2516CK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX BAFF Singleplex Assay Kit is a highly sensitive and quantitative electrochemiluminescent immunoassay for the detection of B cell activating factor (BAFF) in human serum plasma and cell culture supernatants This assay utilizes MSD s proprietary SULFO-TAG labels and multi-array plate technology to enable rapid and accurate measurement of BAFF levels with high sensitivity and a broad dynamic range The assay requires only 50 L of sample and can be read on MSD s SECTOR Imager instruments This assay provides a simple rapid and robust tool for investigating BAFF biology and evaluating BAFF-targeted therapeutics