Learn More
Meso Scale Discovery UPLEX HUM BAFF ASYQP 25 PL
SDP
Catalog No. 5000080611
Encompass
The U-PLEX Human BAFF Assay (K1516CK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of B-cell activating factor (BAFF) in human biological samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable BAFF protein quantification in research applications
Catalog No. 50-000-80611 Supplier Meso Scale Discovery Supplier No. K1516CK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human BAFF Assay (K1516CK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of B-cell activating factor (BAFF) in human biological samples This electrochemiluminescence-based assay utilizes MSD s U-PLEX platform technology offering high sensitivity wide dynamic range and minimal sample volume requirements The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable BAFF protein quantification in research applications