Learn More
Meso Scale Discovery UPLEX HUM BAFFR/TNFRSF13C ASY
SDP
Catalog No. 5000078660
Encompass
The Meso Scale Discovery U-PLEX Human BAFF/TNFRSF13C Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of BAFF and TNFRSF13C in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent It can monitor BAFF and TNFRSF13C levels in only 50 L of sample
Catalog No. 50-000-78660 Supplier Meso Scale Discovery Supplier No. K151K7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human BAFF/TNFRSF13C Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of BAFF and TNFRSF13C in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent It can monitor BAFF and TNFRSF13C levels in only 50 L of sample