Learn More
Meso Scale Discovery UPLEX HUM BAFFR/TNFRSF13C ASY
SDP
Catalog No. 5000079289
Encompass
The Meso Scale Discovery U-PLEX Human BAFF/TNFRSF13C Assay is a multiplex immunoassay kit for measuring BAFF and TNFRSF13C in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label and necessary calibrators and controls for a standard curve and assay validation enabling rapid and accurate quantification from small samples
Catalog No. 50-000-79289 Supplier Meso Scale Discovery Supplier No. K151K7K1
The Meso Scale Discovery U-PLEX Human BAFF/TNFRSF13C Assay is a multiplex immunoassay kit for measuring BAFF and TNFRSF13C in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology offering high sensitivity a dynamic range of up to 5 logs and low sample volume needs The kit includes a 96-well plate with capture antibodies detection antibodies with an electrochemiluminescent label and necessary calibrators and controls for a standard curve and assay validation enabling rapid and accurate quantification from small samples