Learn More
Meso Scale Discovery UPLEX HUM BCMA/TNFRSF17 ASY
SDP
Catalog No. 5000078330
Encompass
The Meso Scale Discovery U-PLEX Human BCMA (TNFRSF17) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of BCMA in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results for BCMA levels The assay has a detection range of 0 156-2500 pg/mL and requires 50 L sample input It can be used to monitor BCMA levels in research and clinical applications involving multiple myeloma and other B-cell malignancies The kit includes a 96-well plate detection antibody calibrator and sample diluent valid for 50 tests when used as directed
Catalog No. 50-000-78330 Supplier Meso Scale Discovery Supplier No. K151D8K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human BCMA (TNFRSF17) Assay is an electrochemiluminescent immunoassay for the quantitative measurement of BCMA in human serum plasma and cell culture supernatants It utilizes MSD s proprietary SULFO-TAG labels and multi-array technology to provide sensitive and reproducible results for BCMA levels The assay has a detection range of 0 156-2500 pg/mL and requires 50 L sample input It can be used to monitor BCMA levels in research and clinical applications involving multiple myeloma and other B-cell malignancies The kit includes a 96-well plate detection antibody calibrator and sample diluent valid for 50 tests when used as directed