Learn More
Meso Scale Discovery UPLEX HUM BCMA/TNFRSF17 ASY384
SDP
Catalog No. 5000078381
Encompass
The Meso Scale Discovery U-PLEX Human BCMA/TNF-RSF17 384-Well Assay Kit is a multiplex immunoassay that measures human BCMA and TNF-RSF17 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and diluent It allows simultaneous quantification of BCMA and TNF-RSF17 from a single small sample volume
Catalog No. 50-000-78381 Supplier Meso Scale Discovery Supplier No. K251D8K2
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human BCMA/TNF-RSF17 384-Well Assay Kit is a multiplex immunoassay that measures human BCMA and TNF-RSF17 in serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample consumption The kit includes a 384-well plate coated with capture antibodies detection antibodies labeled with electrochemiluminescent tags calibrators and diluent It allows simultaneous quantification of BCMA and TNF-RSF17 from a single small sample volume