Learn More
Meso Scale Discovery UPLEX HUM BCMA/TNFRSF17 ASY384
SDP
Catalog No. 5000080820
Encompass
The Meso Scale Discovery BCMA Kit is an electrochemiluminescent immunoassay for measuring BCMA in human serum plasma and cell culture supernatants It uses SULFO-TAG labels and multi-array plate technology for sensitivity and reproducibility The kit includes a 96-well plate with a BCMA capture antibody a detection antibody with a SULFO-TAG label calibrators controls buffers and a plate seal It has a sensitivity of 1-2 pg/mL and a range up to 10 000 pg/mL suitable for monitoring BCMA during immunotherapy
Catalog No. 50-000-80820 Supplier Meso Scale Discovery Supplier No. K251D8K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery BCMA Kit is an electrochemiluminescent immunoassay for measuring BCMA in human serum plasma and cell culture supernatants It uses SULFO-TAG labels and multi-array plate technology for sensitivity and reproducibility The kit includes a 96-well plate with a BCMA capture antibody a detection antibody with a SULFO-TAG label calibrators controls buffers and a plate seal It has a sensitivity of 1-2 pg/mL and a range up to 10 000 pg/mL suitable for monitoring BCMA during immunotherapy