Learn More
Meso Scale Discovery UPLEX HUM BCMA/TNFRSF17 ASYQP
SDP
Catalog No. 5000078742
Encompass
The Meso Scale Discovery U-PLEX Human BCMA Assay is an electrochemiluminescent immunoassay for measuring BCMA in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results in 3 5 hours The assay detects as low as 19 pg/mL and includes a 96-well plate with capture antibodies detection antibodies calibrators and controls It measures soluble BCMA important in B cell maturation and a target for multiple myeloma and other B cell malignancies
Catalog No. 50-000-78742 Supplier Meso Scale Discovery Supplier No. K151D8K22
The Meso Scale Discovery U-PLEX Human BCMA Assay is an electrochemiluminescent immunoassay for measuring BCMA in human serum plasma and cell culture supernatants It uses MSD s SULFO-TAG labels and multi-array technology for sensitive reproducible results in 3 5 hours The assay detects as low as 19 pg/mL and includes a 96-well plate with capture antibodies detection antibodies calibrators and controls It measures soluble BCMA important in B cell maturation and a target for multiple myeloma and other B cell malignancies