Learn More
Meso Scale Discovery UPLEX HUM BCMA/TNFRSF17 ASYQP
SDP
Catalog No. 5000081954
Encompass
The Meso Scale Discovery U-PLEX Human BCMA/TNFRSF17 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of BCMA/TNFRSF17 and up to 14 other human biomarkers It uses MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls It can be used for biomarker research bioanalytical testing and clinical research applications involving human serum plasma and other biological fluids
Catalog No. 50-000-81954 Supplier Meso Scale Discovery Supplier No. K151D8K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human BCMA/TNFRSF17 Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of BCMA/TNFRSF17 and up to 14 other human biomarkers It uses MSD s proprietary electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and controls It can be used for biomarker research bioanalytical testing and clinical research applications involving human serum plasma and other biological fluids