Learn More
Meso Scale Discovery UPLEX HUM BDNF ASYQP 25 PL
SDP
Catalog No. 5000080612
Encompass
The U-PLEX Human BDNF Assay (K1516WK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of brain-derived neurotrophic factor (BDNF) in human biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for BDNF detection in serum plasma and cell culture supernatants The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable reproducible results in neuroscience research applications
Catalog No. 50-000-80612 Supplier Meso Scale Discovery Supplier No. K1516WK24
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human BDNF Assay (K1516WK) is a multiplex immunoassay kit from Meso Scale Discovery for quantitative measurement of brain-derived neurotrophic factor (BDNF) in human biological samples This electrochemiluminescence-based assay offers high sensitivity and specificity for BDNF detection in serum plasma and cell culture supernatants The kit includes pre-coated plates detection antibodies calibrators and all necessary reagents for reliable reproducible results in neuroscience research applications