Learn More
Meso Scale Discovery UPLEX HUM BNGF ASY384well
SDP
Catalog No. 5000080228
Encompass
The Meso Scale Discovery U-PLEX Human Beta-NGF 384-Well Assay is a multiplex immunoassay kit for quantifying beta Nerve Growth Factor (beta-NGF) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology in a 384-well plate format allowing analysis of up to 80 samples per plate The assay has a sensitivity of 0 04 pg/mL and a dynamic range up to 10 000 pg/mL with a total assay time of about 3 5 hours This product aids in understanding beta-NGF biology and evaluating therapeutic interventions targeting beta-NGF pathways
Catalog No. 50-000-80228 Supplier Meso Scale Discovery Supplier No. K2516DK4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human Beta-NGF 384-Well Assay is a multiplex immunoassay kit for quantifying beta Nerve Growth Factor (beta-NGF) in human serum plasma and cell culture supernatants It uses MSD s electrochemiluminescent detection technology in a 384-well plate format allowing analysis of up to 80 samples per plate The assay has a sensitivity of 0 04 pg/mL and a dynamic range up to 10 000 pg/mL with a total assay time of about 3 5 hours This product aids in understanding beta-NGF biology and evaluating therapeutic interventions targeting beta-NGF pathways