Learn More
Meso Scale Discovery UPLEX HUM CA1 ASY384well
SDP
Catalog No. 5000079859
Encompass
The Meso Scale Discovery U-PLEX Human CA1 384-Well Assay is a multiplex immunoassay panel that measures CA1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This 384-well plate assay allows for efficient and high-throughput analysis of CA1 across multiple samples With a short 2 5 hour protocol it enables rapid quantification of CA1 which is involved in pathways related to cancer and immunity This assay requires an MSD instrument such as QuickPlex QuickPlex SQ 120 or QuickPlex SQ 240 for detection
Catalog No. 50-000-79859 Supplier Meso Scale Discovery Supplier No. K251J9K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CA1 384-Well Assay is a multiplex immunoassay panel that measures CA1 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample consumption This 384-well plate assay allows for efficient and high-throughput analysis of CA1 across multiple samples With a short 2 5 hour protocol it enables rapid quantification of CA1 which is involved in pathways related to cancer and immunity This assay requires an MSD instrument such as QuickPlex QuickPlex SQ 120 or QuickPlex SQ 240 for detection