Learn More
Meso Scale Discovery UPLEX HUM CD20 ASY384well
SDP
Catalog No. 5000078375
Encompass
The U-PLEX Human CD20 384-Well Assay from Meso Scale Discovery is a 384-well multiplex immunoassay that measures CD20 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This assay requires only 12 5 L of sample per well and the plate can be read in as little as 2 5 hours It includes a 384-well MULTI-ARRAY Plate pre-coated with capture antibodies detection antibodies and read buffer This assay allows for the quantitative measurement of CD20 levels with accuracy and precision
Catalog No. 50-000-78375 Supplier Meso Scale Discovery Supplier No. K251F7K4
Add to Cart
Add to Cart
Request a Quote
The U-PLEX Human CD20 384-Well Assay from Meso Scale Discovery is a 384-well multiplex immunoassay that measures CD20 in human serum plasma and cell culture supernatants It utilizes electrochemiluminescence detection technology and provides high sensitivity broad dynamic range and rapid results This assay requires only 12 5 L of sample per well and the plate can be read in as little as 2 5 hours It includes a 384-well MULTI-ARRAY Plate pre-coated with capture antibodies detection antibodies and read buffer This assay allows for the quantitative measurement of CD20 levels with accuracy and precision