Learn More
Meso Scale Discovery UPLEX HUM CD20 ASYQP 25 PL
SDP
Catalog No. 5000078536
Encompass
The Meso Scale Discovery U-PLEX Human CD20 Assay Kit is a multiplex immunoassay for detecting and quantifying CD20 and up to seven other biomarkers from one sample It uses electrochemiluminescence (ECL) technology on the QuickPlex SQ 120 instrument for high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibodies ruthenium-labeled detection antibodies calibrator and sample diluent It requires 25 L of sample per well and works with serum plasma or cell culture supernatants with a detection limit of 0 07 ng/mL for CD20
Catalog No. 50-000-78536 Supplier Meso Scale Discovery Supplier No. K151F7K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CD20 Assay Kit is a multiplex immunoassay for detecting and quantifying CD20 and up to seven other biomarkers from one sample It uses electrochemiluminescence (ECL) technology on the QuickPlex SQ 120 instrument for high sensitivity and a broad dynamic range The kit includes a 96-well plate with capture antibodies ruthenium-labeled detection antibodies calibrator and sample diluent It requires 25 L of sample per well and works with serum plasma or cell culture supernatants with a detection limit of 0 07 ng/mL for CD20