Learn More
Meso Scale Discovery UPLEX HUM CD27 ASYQP 25 PL
SDP
Catalog No. 5000082009
Encompass
The Meso Scale Discovery U-PLEX Human CD27 Assay Kit is a multiplex immunoassay that measures CD27 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD Read Buffer It can test up to 80 samples in duplicate and has a testing time of 3 5 hours The kit has been optimized and validated for research use only
Catalog No. 50-000-82009 Supplier Meso Scale Discovery Supplier No. K151G7K24
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CD27 Assay Kit is a multiplex immunoassay that measures CD27 in human serum plasma and cell culture supernatants It uses electrochemiluminescence detection technology and provides high sensitivity up to 5-logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and an MSD Read Buffer It can test up to 80 samples in duplicate and has a testing time of 3 5 hours The kit has been optimized and validated for research use only