Learn More
Meso Scale Discovery UPLEX HUM CD276/B7H3 ASY
SDP
Catalog No. 5000081634
Encompass
The Meso Scale Discovery U-PLEX Human CD276 (B7-H3) Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of CD276 and other biomarkers in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent It can monitor CD276 levels and has research applications in oncology immunology and other life science fields
Catalog No. 50-000-81634 Supplier Meso Scale Discovery Supplier No. K151E7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery U-PLEX Human CD276 (B7-H3) Assay is a multiplex immunoassay kit that enables simultaneous quantitative measurement of CD276 and other biomarkers in human serum plasma and cell culture supernatants It utilizes MSD s electrochemiluminescent detection technology and provides high sensitivity up to 5 logs of dynamic range and low sample volume requirements This assay kit includes a 96-well plate coated with capture antibodies detection antibodies labeled with MSD SULFO-TAG calibrators and diluent It can monitor CD276 levels and has research applications in oncology immunology and other life science fields