Learn More
Meso Scale Discovery UPLEX HUM CD276/B7H3 ASY384
SDP
Catalog No. 5000080993
Encompass
The Meso Scale Discovery CD276 (B7-H3) Kit is an electrochemiluminescent immunoassay for measuring CD276 (B7-H3) in human serum plasma and cell culture supernatants It includes a 96-well plate with a CD276 capture antibody a detection antibody with an electrochemiluminescent label calibrator diluent wash buffer read buffer and lyophilized calibrator The assay sensitivity is 0 03 ng/mL with a range of 0 03 to 400 ng/mL Utilizing MSD s MULTI-ARRAY technology it offers high sensitivity specificity and reproducibility with minimal cross-reactivity
Catalog No. 50-000-80993 Supplier Meso Scale Discovery Supplier No. K251E7K4
Add to Cart
Add to Cart
Request a Quote
The Meso Scale Discovery CD276 (B7-H3) Kit is an electrochemiluminescent immunoassay for measuring CD276 (B7-H3) in human serum plasma and cell culture supernatants It includes a 96-well plate with a CD276 capture antibody a detection antibody with an electrochemiluminescent label calibrator diluent wash buffer read buffer and lyophilized calibrator The assay sensitivity is 0 03 ng/mL with a range of 0 03 to 400 ng/mL Utilizing MSD s MULTI-ARRAY technology it offers high sensitivity specificity and reproducibility with minimal cross-reactivity